siRNA Details
| Gene Symbol: Rdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765053.1 | XP_008763275.1 | 3 | 899 | ||
| XM_008765054.1 | XP_008763276.1 | 3 | 894 | |||
| XM_008765055.1 | XP_008763277.1 | 3 | 883 | |||
| XM_008765056.1 | XP_008763278.1 | 3 | 899 | |||
| GenBank mRNA | FQ218828.1 | - | 3 | 466 | ||

Rdh5
Genomic Map