siRNA Details
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108989.1 | NP_001102459.1 | 3 | 1269 | ||
XM_008765202.1 | XP_008763424.1 | 3 | 1296 | |||
GenBank mRNA | BC166847.1 | - | 3 | 1252 | 9030846 |
