siRNA Details
| Gene Symbol: Grap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034944.1 | NP_001030116.1 | 4 | 505 | ||
| GenBank mRNA | BC105895.1 | - | 4 | 505 | 7380480 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grap2| Gene Symbol: Grap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034944.1 | NP_001030116.1 | 4 | 505 | ||
| GenBank mRNA | BC105895.1 | - | 4 | 505 | 7380480 | |