siRNA Details
| Gene Symbol: Ap1m2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108996.2 | NP_001102466.1 | 1 | 54 | ||
| GenBank mRNA | BC167082.1 | - | 1 | 54 | 7930480 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap1m2| Gene Symbol: Ap1m2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108996.2 | NP_001102466.1 | 1 | 54 | ||
| GenBank mRNA | BC167082.1 | - | 1 | 54 | 7930480 | |