siRNA Details
Gene Symbol: Olr1230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000595.1 | NP_001000595.1 | 1 | 799 | ||
XM_008762652.1 | XP_008760874.1 | 3 | 989 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000595.1 | NP_001000595.1 | 1 | 799 | ||
XM_008762652.1 | XP_008760874.1 | 3 | 989 |