siRNA Details
| Gene Symbol: Neil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025754.1 | NP_001020925.1 | 2 | 337 | ||
| XM_006243134.2 | XP_006243196.1 | 2 | 1151 | |||
| XM_006243135.2 | XP_006243197.1 | 2 | 1257 | |||
| XM_006243136.2 | XP_006243198.1 | 3 | 333 | |||
| GenBank mRNA | BC099125.1 | - | 2 | 337 | 7445605 | |

Neil1
Genomic Map