siRNA Details
| Gene Symbol: Foxb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013248.1 | NP_001013266.1 | 2 | 341 | ||
| GenBank mRNA | BC089975.1 | - | 2 | 341 | 7316995 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxb1| Gene Symbol: Foxb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013248.1 | NP_001013266.1 | 2 | 341 | ||
| GenBank mRNA | BC089975.1 | - | 2 | 341 | 7316995 | |