siRNA Details
| Gene Symbol: Hmgcll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276472.1 | NP_001263401.1 | 4 | 441 | ||
| XM_006243415.2 | XP_006243477.1 | 4 | 1110 | |||
| GenBank mRNA | FQ211656.1 | - | 4 | 441 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmgcll1| Gene Symbol: Hmgcll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276472.1 | NP_001263401.1 | 4 | 441 | ||
| XM_006243415.2 | XP_006243477.1 | 4 | 1110 | |||
| GenBank mRNA | FQ211656.1 | - | 4 | 441 | ||