siRNA Details
| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109007.2 | NP_001102477.2 | 19 | 2446 | ||
| NM_001172159.1 | NP_001165630.1 | 18 | 2404 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dis3l2| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109007.2 | NP_001102477.2 | 19 | 2446 | ||
| NM_001172159.1 | NP_001165630.1 | 18 | 2404 | |||