siRNA Details
| Gene Symbol: Lrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009717.1 | NP_001009717.1 | 2 | 681 | ||
| GenBank mRNA | BC088434.1 | - | 2 | 681 | 7124833 | |
| FQ220477.1 | - | 2 | 691 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrg1| Gene Symbol: Lrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009717.1 | NP_001009717.1 | 2 | 681 | ||
| GenBank mRNA | BC088434.1 | - | 2 | 681 | 7124833 | |
| FQ220477.1 | - | 2 | 691 | |||