siRNA Details
| Gene Symbol: Praf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109013.1 | NP_001102483.1 | 3 | 613 | ||
| GenBank mRNA | FQ212649.1 | - | 3 | 472 | ||
| FQ213026.1 | - | 3 | 472 | |||
| FQ213646.1 | - | 3 | 469 | |||
| FQ219908.1 | - | 3 | 472 | |||

Praf2
Genomic Map