siRNA Details
| Gene Symbol: Asb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191913.1 | NP_001178842.1 | 5 | 468 | ||
| XM_006256881.2 | XP_006256943.1 | 6 | 809 | |||
| XM_006256883.2 | XP_006256945.1 | 6 | 650 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb9| Gene Symbol: Asb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191913.1 | NP_001178842.1 | 5 | 468 | ||
| XM_006256881.2 | XP_006256943.1 | 6 | 809 | |||
| XM_006256883.2 | XP_006256945.1 | 6 | 650 | |||