siRNA Details
| Gene Symbol: Bmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109016.1 | NP_001102486.1 | 11, 12 | 1220 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmx| Gene Symbol: Bmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109016.1 | NP_001102486.1 | 11, 12 | 1220 | ||