siRNA Details
| Gene Symbol: Zfx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109017.1 | NP_001102487.1 | 4 | 959 | ||
| XM_006257030.2 | XP_006257092.1 | 4 | 682 | |||
| XM_006257031.2 | XP_006257093.1 | 3 | 962 | |||
| XM_006257032.2 | XP_006257094.1 | 4 | 1029 | |||
| XM_008773264.1 | XP_008771486.1 | 3 | 614 | |||
| XM_008773265.1 | XP_008771487.1 | 4 | 1063 | |||
| XM_008773266.1 | XP_008771488.1 | 3 | 614 | |||

Zfx
Genomic Map