siRNA Details
| Gene Symbol: Btaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191917.1 | NP_001178846.1 | 19 | 2341 | ||
| XM_006231332.2 | XP_006231394.1 | 19 | 2627 | |||
| XM_006231334.1 | XP_006231396.1 | 16 | 2003 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btaf1| Gene Symbol: Btaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191917.1 | NP_001178846.1 | 19 | 2341 | ||
| XM_006231332.2 | XP_006231394.1 | 19 | 2627 | |||
| XM_006231334.1 | XP_006231396.1 | 16 | 2003 | |||