siRNA Details
| Gene Symbol: Atp6v1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212490.2 | NP_997655.1 | 1 | 257 | ||
| GenBank mRNA | FQ212526.1 | - | 1 | 186 | ||
| FQ212959.1 | - | 1 | 219 | |||
| FQ213756.1 | - | 1 | 110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v1g2| Gene Symbol: Atp6v1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212490.2 | NP_997655.1 | 1 | 257 | ||
| GenBank mRNA | FQ212526.1 | - | 1 | 186 | ||
| FQ212959.1 | - | 1 | 219 | |||
| FQ213756.1 | - | 1 | 110 | |||