siRNA Details
| Gene Symbol: Dnajc30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109024.2 | NP_001102494.1 | 1 | 357 | ||
| GenBank mRNA | BC166911.1 | - | 1 | 357 | 6890506 | |
| FQ214485.1 | - | 1 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc30| Gene Symbol: Dnajc30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109024.2 | NP_001102494.1 | 1 | 357 | ||
| GenBank mRNA | BC166911.1 | - | 1 | 357 | 6890506 | |
| FQ214485.1 | - | 1 | 406 | |||