siRNA Details
| Gene Symbol: Krt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183333.1 | NP_899162.1 | 1013 | |||
| GenBank mRNA | AY342389.1 | - | 1013 | |||
| BC062086.1 | - | 876 | 5599969 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt5| Gene Symbol: Krt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183333.1 | NP_899162.1 | 1013 | |||
| GenBank mRNA | AY342389.1 | - | 1013 | |||
| BC062086.1 | - | 876 | 5599969 | |||