siRNA Details
| Gene Symbol: Krt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183333.1 | NP_899162.1 | 460 | |||
| GenBank mRNA | AY342389.1 | - | 460 | |||
| BC062086.1 | - | 323 | 5599969 | |||
| Gene Symbol: LOC102551453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765863.1 | XP_008764085.1 | 1 | 348 | ||

Krt5
Genomic Map