siRNA Details
| Gene Symbol: Il18rap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767060.1 | XP_008765282.1 | 3, 4 | 1043 | ||
| GenBank mRNA | AJ550893.1 | - | 1, 2 | 60 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il18rap| Gene Symbol: Il18rap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767060.1 | XP_008765282.1 | 3, 4 | 1043 | ||
| GenBank mRNA | AJ550893.1 | - | 1, 2 | 60 | ||