siRNA Details
| Gene Symbol: Stk25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_184049.1 | NP_908938.1 | 7 | 904 | ||
| GenBank mRNA | AY346152.1 | - | 7 | 904 | ||
| BC087092.1 | - | 7 | 850 | 7190997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk25| Gene Symbol: Stk25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_184049.1 | NP_908938.1 | 7 | 904 | ||
| GenBank mRNA | AY346152.1 | - | 7 | 904 | ||
| BC087092.1 | - | 7 | 850 | 7190997 | ||