siRNA Details
Gene Symbol: Pcdha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199503.1 | NP_955797.1 | 1 | 754 | ||
GenBank mRNA | AB113384.1 | - | 1 | 754 | ||
AY573971.1 | - | 1 | 794 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pcdha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199503.1 | NP_955797.1 | 1 | 754 | ||
GenBank mRNA | AB113384.1 | - | 1 | 754 | ||
AY573971.1 | - | 1 | 794 |