siRNA Details
| Gene Symbol: Pcdha6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199506.1 | NP_955800.1 | 1 | 1816 | ||
| GenBank mRNA | AB113389.1 | - | 1 | 1816 | ||
| AY573980.1 | - | 1 | 1765 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdha6| Gene Symbol: Pcdha6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199506.1 | NP_955800.1 | 1 | 1816 | ||
| GenBank mRNA | AB113389.1 | - | 1 | 1816 | ||
| AY573980.1 | - | 1 | 1765 | |||