siRNA Details
| Gene Symbol: Pcdha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199507.1 | NP_955801.1 | 1 | 1165 | ||
| GenBank mRNA | AB113390.1 | - | 1 | 1165 | ||
| AY573981.1 | - | 1 | 1165 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdha7| Gene Symbol: Pcdha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199507.1 | NP_955801.1 | 1 | 1165 | ||
| GenBank mRNA | AB113390.1 | - | 1 | 1165 | ||
| AY573981.1 | - | 1 | 1165 | |||