siRNA Details
Gene Symbol: Pcdha9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199508.1 | NP_955802.1 | 1 | 2139 | ||
GenBank mRNA | AB113392.1 | - | 1 | 2139 | ||
AY573983.1 | - | 1 | 2139 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pcdha9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199508.1 | NP_955802.1 | 1 | 2139 | ||
GenBank mRNA | AB113392.1 | - | 1 | 2139 | ||
AY573983.1 | - | 1 | 2139 |