siRNA Details
| Gene Symbol: Pcdhac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199509.1 | NP_955803.1 | 1 | 1858 | ||
| GenBank mRNA | AB113397.1 | - | 1 | 1858 | ||
| AY573984.1 | - | 1 | 1858 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhac1| Gene Symbol: Pcdhac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199509.1 | NP_955803.1 | 1 | 1858 | ||
| GenBank mRNA | AB113397.1 | - | 1 | 1858 | ||
| AY573984.1 | - | 1 | 1858 | |||