siRNA Details
| Gene Symbol: Adm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201426.1 | NP_958829.1 | 3 | 343 | ||
| GenBank mRNA | AB121036.1 | - | 3 | 343 | ||
| AB181297.1 | - | 3 | 352 | |||
| AY590103.1 | - | 2 | 171 | |||
| FQ234235.1 | - | 3 | 336 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adm2| Gene Symbol: Adm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201426.1 | NP_958829.1 | 3 | 343 | ||
| GenBank mRNA | AB121036.1 | - | 3 | 343 | ||
| AB181297.1 | - | 3 | 352 | |||
| AY590103.1 | - | 2 | 171 | |||
| FQ234235.1 | - | 3 | 336 | |||