siRNA Details
| Gene Symbol: Mrgprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002281.1 | NP_001002281.1 | 1 | 936 | ||
| XM_006227882.2 | XP_006227944.1 | 2 | 1188 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrgprh| Gene Symbol: Mrgprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002281.1 | NP_001002281.1 | 1 | 936 | ||
| XM_006227882.2 | XP_006227944.1 | 2 | 1188 | |||
Set Membership: |
|