siRNA Details
| Gene Symbol: Olr1353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000750.1 | NP_001000750.1 | 1 | 834 | ||
| XM_006245545.2 | XP_006245607.1 | 4 | 1324 | |||
| XM_006245546.2 | XP_006245608.1 | 3 | 1176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1353| Gene Symbol: Olr1353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000750.1 | NP_001000750.1 | 1 | 834 | ||
| XM_006245545.2 | XP_006245607.1 | 4 | 1324 | |||
| XM_006245546.2 | XP_006245608.1 | 3 | 1176 | |||