siRNA Details
| Gene Symbol: Olr1303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000796.1 | NP_001000796.1 | 1 | 106 | ||
| Gene Symbol: LOC100910667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766112.1 | XP_008764334.1 | 106 | |||

Olr1303
Genomic Map