siRNA Details
| Gene Symbol: Olr491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000932.1 | NP_001000932.1 | 906 | |||
| Gene Symbol: LOC100909831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749506.1 | XP_003749554.1 | 1 | 906 | ||

Olr491
Genomic Map