siRNA Details
| Gene Symbol: Olr1474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000943.1 | NP_001000943.1 | 1 | 795 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1474| Gene Symbol: Olr1474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000943.1 | NP_001000943.1 | 1 | 795 | ||