siRNA Details
| Gene Symbol: Olr1445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000958.1 | NP_001000958.1 | 1 | 447 | ||
| Gene Symbol: LOC100911586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750835.1 | XP_003750883.1 | 447 | |||

Olr1445
Genomic Map