siRNA Details
Gene Symbol: Olr1615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001003.1 | NP_001001003.1 | 1 | 617 | ||
XM_006251847.2 | XP_006251909.1 | 2 | 646 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
