siRNA Details
| Gene Symbol: Olr51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001021.1 | NP_001001021.1 | 1 | 936 | ||
| Gene Symbol: LOC100912515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229824.1 | XP_006229886.1 | 1 | 936 | ||

Olr51
Genomic Map