siRNA Details
| Gene Symbol: Olr1397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001094.1 | NP_001001094.1 | 1 | 13 | ||
| XM_008767693.1 | XP_008765915.1 | 4 | 1048 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1397| Gene Symbol: Olr1397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001094.1 | NP_001001094.1 | 1 | 13 | ||
| XM_008767693.1 | XP_008765915.1 | 4 | 1048 | |||