siRNA Details
Gene Symbol: Olr1528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001099.1 | NP_001001099.1 | 1 | 476 | ||
XM_006248194.2 | XP_006248256.1 | 2 | 844 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001099.1 | NP_001001099.1 | 1 | 476 | ||
XM_006248194.2 | XP_006248256.1 | 2 | 844 |