siRNA Details
| Gene Symbol: Olr1529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001100.1 | NP_001001100.1 | 1 | 577 | ||
| XM_006248195.2 | XP_006248257.1 | 2 | 676 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1529| Gene Symbol: Olr1529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001100.1 | NP_001001100.1 | 1 | 577 | ||
| XM_006248195.2 | XP_006248257.1 | 2 | 676 | |||