siRNA Details
| Gene Symbol: Olr1530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001101.1 | NP_001001101.1 | 1 | 665 | ||
| GenBank mRNA | AF091575.1 | - | 1 | 479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1530| Gene Symbol: Olr1530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001101.1 | NP_001001101.1 | 1 | 665 | ||
| GenBank mRNA | AF091575.1 | - | 1 | 479 | ||