siRNA Details
| Gene Symbol: Gpsm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003974.2 | NP_001003974.1 | 1, 2 | 350 | ||
| GenBank mRNA | BC088220.1 | - | 1, 2 | 350 | 7384543 | |
| FQ229559.1 | - | 1, 2 | 170 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpsm3| Gene Symbol: Gpsm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003974.2 | NP_001003974.1 | 1, 2 | 350 | ||
| GenBank mRNA | BC088220.1 | - | 1, 2 | 350 | 7384543 | |
| FQ229559.1 | - | 1, 2 | 170 | |||