siRNA Details
| Gene Symbol: Egfl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004070.2 | NP_001004070.1 | 5, 6 | 592 | ||
| NM_001165880.1 | NP_001159352.1 | 6, 7 | 740 | |||
| XM_006255977.2 | XP_006256039.1 | 6, 7 | 1461 | |||
| XM_006255978.2 | XP_006256040.1 | 7, 8 | 1347 | |||
| XM_006255979.2 | XP_006256041.1 | 7, 8 | 843 | |||
| GenBank mRNA | BC127463.1 | - | 6, 7 | 740 | 8362353 | |

Egfl8
Genomic Map