siRNA Details
Gene Symbol: Krt77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008807.1 | NP_001008807.1 | 1 | 175 | ||
XM_006242435.2 | XP_006242497.1 | 1 | 318 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krt77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008807.1 | NP_001008807.1 | 1 | 175 | ||
XM_006242435.2 | XP_006242497.1 | 1 | 318 |