siRNA Details
| Gene Symbol: Clic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002807.2 | NP_001002807.1 | 4, 5 | 610 | ||
| XM_008772745.1 | XP_008770967.1 | 4, 5 | 611 | |||
| GenBank mRNA | BC099823.1 | - | 4, 5 | 562 | 7932557 | |
| EU148064.1 | - | 4, 5 | 373 | |||
| FM115459.1 | - | 2, 3 | 232 | |||
| FQ219968.1 | - | 4, 5 | 600 | |||
| FQ227136.1 | - | 4, 5 | 396 | |||
| FQ227567.1 | - | 4, 5 | 387 | |||
| FQ227951.1 | - | 4, 5 | 387 | |||
| FQ233892.1 | - | 4, 5 | 387 | |||

Clic1
Genomic Map