siRNA Details
| Gene Symbol: Clic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002807.2 | NP_001002807.1 | 5 | 643 | ||
| XM_008772745.1 | XP_008770967.1 | 5 | 644 | |||
| GenBank mRNA | EU148064.1 | - | 5 | 406 | ||
| FM115459.1 | - | 3 | 265 | |||
| FQ219968.1 | - | 5 | 633 | |||
| FQ227136.1 | - | 5 | 429 | |||
| FQ227567.1 | - | 5 | 420 | |||
| FQ227951.1 | - | 5 | 420 | |||
| FQ233892.1 | - | 5 | 420 | |||

Clic1
Genomic Map