siRNA Details
| Gene Symbol: Krt26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008823.1 | NP_001008823.1 | 5 | 859 | ||
| XM_006247390.2 | XP_006247452.1 | 5 | 1447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt26| Gene Symbol: Krt26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008823.1 | NP_001008823.1 | 5 | 859 | ||
| XM_006247390.2 | XP_006247452.1 | 5 | 1447 | |||