siRNA Details
| Gene Symbol: Krt85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008811.1 | NP_001008811.1 | 3 | 672 | ||
| XM_006242351.2 | XP_006242413.1 | 3 | 728 | |||
| GenBank mRNA | AB355640.1 | - | 3 | 672 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt85| Gene Symbol: Krt85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008811.1 | NP_001008811.1 | 3 | 672 | ||
| XM_006242351.2 | XP_006242413.1 | 3 | 728 | |||
| GenBank mRNA | AB355640.1 | - | 3 | 672 | ||