siRNA Details
| Gene Symbol: St6galnac6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015036.1 | NP_001015036.1 | 4 | 951 | ||
| XM_006233982.2 | XP_006234044.1 | 5 | 1390 | |||
| XM_006233984.2 | XP_006234046.1 | 6 | 1557 | |||
| XM_006233985.2 | XP_006234047.1 | 5 | 1392 | |||
| XM_006233987.2 | XP_006234049.1 | 6 | 1022 | |||
| XM_006233988.2 | XP_006234050.1 | 6 | 1026 | |||
| XM_008761770.1 | XP_008759992.1 | 7 | 1234 | |||
| XM_008761771.1 | XP_008759993.1 | 7 | 1237 | |||
| GenBank mRNA | AJ646881.1 | - | 5 | 835 | ||
| BC091354.1 | - | 4 | 951 | 7317530 | ||

St6galnac6
Genomic Map