siRNA Details
| Gene Symbol: Rnf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109025.2 | NP_001102495.1 | 1 | 192 | ||
| GenBank mRNA | BC088341.1 | - | 1 | 168 | 7367801 | |
| DY573537.1 | - | 1 | 192 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf5| Gene Symbol: Rnf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109025.2 | NP_001102495.1 | 1 | 192 | ||
| GenBank mRNA | BC088341.1 | - | 1 | 168 | 7367801 | |
| DY573537.1 | - | 1 | 192 | |||