siRNA Details
Gene Symbol: Wfdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008868.1 | NP_001008868.1 | 2 | 207 | ||
XM_008762531.1 | XP_008760753.1 | 2 | 218 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wfdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008868.1 | NP_001008868.1 | 2 | 207 | ||
XM_008762531.1 | XP_008760753.1 | 2 | 218 |