siRNA Details
| Gene Symbol: Spink14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008875.1 | NP_001008875.1 | 3 | 171 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spink14| Gene Symbol: Spink14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008875.1 | NP_001008875.1 | 3 | 171 | ||